elements™ Modular Magnetic Stirrer
Actual product may vary
Cole-Parmer
elements™ Modular Magnetic Stirrer
Optimize your stirring needs and workspace with this completely customizable stirrer
Have Questions?
Catalog NumberTypeStirring Positions
CLP84003-81Stirrer Control Unit10
Catalog number CLP84003-81
Price (MYR)
-
Type:
Stirrer Control Unit
Stirring Positions:
10
  • Attach up to 10 stir plates based on need
  • Simple touch and slide operation
  • LCD display with real time parameters
  • Corrosion-resistant glass stir top
  • Waterproof USB attachments
Specifications
Speed100 to 1500 rpm
Stirring Positions10
Top Plate MaterialGlass
Capacity1 L/position
DisplayDigital
ShapeRound
Voltage100/240 V
Frequency50/60 Hz
TypeStirrer Control Unit
Unit SizeEach