QuickConnect™ Mop Head Frames
Actual product may vary
Masterflex
QuickConnect™ Mop Head Frames
Featuring a quick-connect button for easy attachment to all sizes and styles of Contec™ QuickConnect™ handles. Contec™ QuickConnect™ Mop Head Frames are specifically designed for use with the Contec™ family of mopping systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Length (Metric)
CLP40614-38Medium and Large VertiKlean MAX Mop Heads29 cm
CTC2652SSMedium and Large VertiKlean MAX Mop Heads29 cm
Catalog number CLP40614-38
Price (MYR)
-
For Use With (Equipment):
Medium and Large VertiKlean MAX Mop Heads
Length (Metric):
29 cm
  • Offer maximum flexibility for cleaning in critical environments.
  • Interchangeable frames for Contec Mopping Systems
  • Autoclavable
Specifications
Color CodeWhite
Product TypeMop Head Frame
Length (English)11.5 in.
SterilityNon-sterile
Length (Metric)29 cm
AutoclavableAutoclavable
DisposableReusable
For Use With (Equipment)Medium and Large VertiKlean MAX Mop Heads
For Use With (Application)Pharmaceutical and medical device manufacturing facilities, or other similar critical or controlled environments
Color-codedNo