Product Image
Acros Organics
Poly(ethylene glycol) bis(3-aminopropyl) terminated
CAS: 34901-14-9 | (C2H4O)nC6H16N2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number ACR460610010
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPoly(ethylene glycol) bis(3-aminopropyl) terminated
Melting Point51.5°C to 54.0°C
CAS34901-14-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement Causes skin irritation.