Acetic acid, 25%, solution in water
Acetic acid, 25%, solution in water
Acetic acid, 25%, solution in water
Thermo Scientific Chemicals
Acetic acid, 25%, solution in water
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
Catalog number ACR456850050
Price (MYR)
557.00
Each
Quantity:
5 L
Request bulk or custom format
Price (MYR)
557.00
Each
Specifications
Chemical Name or MaterialAcetic acid
CAS64-19-7
Melting Point-17.0°C
Boiling Point103.0°C
Health Hazard 1Danger