3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
Acros Organics
3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%
CAS: 2421-28-5 | C17H6O7 | 322.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
Glass bottle
Catalog number ACR401930050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3,3′,4,4′-Benzophenonetetracarboxylic dianhydride
Name Note99%
CAS2421-28-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning