Diphenhydramine hydrochloride, 99%
Diphenhydramine hydrochloride, 99%
Diphenhydramine hydrochloride, 99%
Diphenhydramine hydrochloride, 99%
Thermo Scientific Chemicals
Diphenhydramine hydrochloride, 99%
CAS: 147-24-0 | C17H22ClNO | 291.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR350851000
Price (MYR)
734.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
734.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiphenhydramine hydrochloride
Name Note99%
CAS147-24-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: