O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%
O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%
O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%
Acros Organics
O-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 98%
CAS: 94790-37-1 | C11H16F6N5OP | 379.247 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR299430010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialO-(Benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate
CAS94790-37-1
Assay Percent Range98%
PackagingGlass bottle
Molecular FormulaC11H16F6N5OP
View more
Coupling reagent
RUO – Research Use Only