6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific™
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific™
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific™
Acros Organics
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR292980050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
CAS2328-12-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only