Methyltrimethoxysilane, 97%
Methyltrimethoxysilane, 97%
Methyltrimethoxysilane, 97%
Acros Organics
Methyltrimethoxysilane, 97%
CAS: 1185-55-3 | C4H12O3Si | 136.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR25054050050 mLGlass bottle
Catalog number ACR250540500
Price (MYR)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyltrimethoxysilane
Name Note97%
CAS1185-55-3
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger