2-Iodoanisole, 97%
2-Iodoanisole, 97%
2-Iodoanisole, 97%
Acros Organics
2-Iodoanisole, 97%
CAS: 529-28-2 | C7H7IO | 234.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
Packaging:
Glass bottle
Catalog number ACR249680500
Price (MYR)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Iodoanisole
Name Note97%
CAS529-28-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning