Inosine 5'-monophosphate, disodium salt hydrate, 97%
Inosine 5'-monophosphate, disodium salt hydrate, 97%
Inosine 5'-monophosphate, disodium salt hydrate, 97%
Inosine 5'-monophosphate, disodium salt hydrate, 97%
Thermo Scientific Chemicals
Inosine 5'-monophosphate, disodium salt hydrate, 97%
CAS: 352195-40-5 | C10H11N4Na2O8P | 392.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR226261000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialInosine 5'-monophosphate, disodium salt hydrate
CAS352195-40-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Health Hazard 3GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.