Carnauba wax, refined, No. 1, yellow
Thermo Scientific Chemicals
Carnauba wax, refined, No. 1, yellow
CAS: 8015-86-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Plastic bottle
Catalog number ACR222380010
Price (MYR)
1,001.00
Bottle
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,001.00
Bottle
Specifications
Chemical Name or MaterialCarnauba wax
Acidity2 to 7 (mg KOH/g)
CAS8015-86-9
Melting Point81.0°C to 86.0°C
ColorYellow