Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Triton™ X-100, Thermo Scientific™
Acros Organics
Triton™ X-100, Thermo Scientific™
Have Questions?
Catalog NumberQuantityPackaging
ACR215682500250 mLGlass Bottle
Catalog number ACR215682500
Price (MYR)
-
Quantity:
250 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTriton™ X-100
CAS9002-93-1
Health Hazard 1Danger
Boiling Point270.0°C
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects.