Cyanamide, 95%, stabilized
Cyanamide, 95%, stabilized
Cyanamide, 95%, stabilized
Acros Organics
Cyanamide, 95%, stabilized
CAS: 420-04-2 | CH2N2 | 42.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR181951000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCyanamide
Name Note95%
CAS420-04-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic in contact with skin.
Suspecte