Acrylic acid, 98%, extra pure, stabilized
Acrylic acid, 98%, extra pure, stabilized
Acrylic acid, 98%, extra pure, stabilized
Thermo Scientific Chemicals
Acrylic acid, 98%, extra pure, stabilized
CAS: 79-10-7 | C3H4O2 | 72.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR164250025
Price (MYR)
921.00
Bottle
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
921.00
Bottle
Specifications
Chemical Name or MaterialAcrylic acid
Name NoteStabilized
CAS79-10-7
CAS Min %98
Health Hazard 1GHS Signal Word: Danger