▸
Organic compounds
▸
Organoheterocyclic compounds
▸
Indoles and derivatives
▸
5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™
Element Search
Structure Search
Acros Organics
5-Methoxytryptamine hydrochloride, 97%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
ACR126130010
1 g
Glass bottle
Catalog number
ACR126130010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
5-Methoxytryptamine hydrochloride
Name Note
97%
CAS
66-83-1
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Warning