3'-Methoxyacetophenone, 98%
3'-Methoxyacetophenone, 98%
3'-Methoxyacetophenone, 98%
Acros Organics
3'-Methoxyacetophenone, 98%
CAS: 586-37-8 | C9H10O2 | 150.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR12568050050 mLGlass bottle
Catalog number ACR125680500
Price (MYR)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3'-Methoxyacetophenone
Name Note98%
CAS586-37-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning