Melamine, 99%
Melamine, 99%
Melamine, 99%
Thermo Scientific Chemicals
Melamine, 99%
CAS: 108-78-1 | C3H6N6 | 126.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
1 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number ACR125350050
Price (MYR)
396.00
Bottle
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
396.00
Bottle
Specifications
Chemical Name or MaterialMelamine
CAS108-78-1
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Melting Point354°C