Lithium tri-tert-butoxyaluminohydride, 93-98%
Lithium tri-tert-butoxyaluminohydride, 93-98%
Lithium tri-tert-butoxyaluminohydride, 93-98%
Acros Organics
Lithium tri-tert-butoxyaluminohydride, 93-98%
CAS: 17476-04-9 | C12H28AlLiO3 | 254.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number ACR125160250
Price (MYR)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLithium tri-tert-butoxyaluminohydride
Name Note93-98%
CAS17476-04-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
View more
Reduction reagent
RUO – Research Use Only