1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
Acros Organics
1-(2-Chloroethyl)-4-methoxybenzene, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
Packaging:
Glass bottle
Catalog number ACR123111000
Price (MYR)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-(2-Chloroethyl)-4-methoxybenzene
Name Note97%
CAS18217-00-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning