5-Hydroxy-2-nitrobenzaldehyde, 98%
5-Hydroxy-2-nitrobenzaldehyde, 98%
5-Hydroxy-2-nitrobenzaldehyde, 98%
Acros Organics
5-Hydroxy-2-nitrobenzaldehyde, 98%
CAS: 42454-06-8 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number ACR121650050
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material5-Hydroxy-2-nitrobenzaldehyde
CAS42454-06-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a