(-)-Di-p-toluoyl-L-tartaric acid, 97%
(-)-Di-p-toluoyl-L-tartaric acid, 97%
(-)-Di-p-toluoyl-L-tartaric acid, 97%
Acros Organics
(-)-Di-p-toluoyl-L-tartaric acid, 97%
CAS: 32634-66-5 | C20H18O8 | 386.36 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ACR117571000100 gGlass bottle
Catalog number ACR117571000
Price (MYR)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(-)-Di-p-toluoyl-L-tartaric acid
CAS32634-66-5
CAS Min %96.0
CAS Max %100.0
Melting Point164°C to 170°C