4-Bromopyridine Hydrochloride, 98%
4-Bromopyridine Hydrochloride, 98%
4-Bromopyridine Hydrochloride, 98%
Acros Organics
4-Bromopyridine Hydrochloride, 98%
CAS: 19524-06-2 | C5H4BrN·HCl | 194.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number ACR107420010
Price (MYR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromopyridine hydrochloride
CAS19524-06-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0