1-Bromoheptane, 99%
1-Bromoheptane, 99%
1-Bromoheptane, 99%
Acros Organics
1-Bromoheptane, 99%
CAS: 629-04-9 | C7H15Br | 179.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number ACR107030050
Price (MYR)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromoheptane
Name Note99%
CAS629-04-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning