L(+)-Arginine Hydrochloride, 98+%, Thermo Scientific Chemicals
L(+)-Arginine Hydrochloride, 98+%, Thermo Scientific Chemicals
L(+)-Arginine Hydrochloride, 98+%, Thermo Scientific Chemicals
Acros Organics
L(+)-Arginine Hydrochloride, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Plastic bottle
Catalog number ACR105001000
Price (MYR)
462.00
Bottle
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
462.00
Bottle
Specifications
Chemical Name or MaterialL(+)-Arginine hydrochloride
CAS1119-34-2
Melting Point226°C
ColorWhite
EINECS Number214-275-1