4-Aminophenyl ether, 98%
4-Aminophenyl ether, 98%
4-Aminophenyl ether, 98%
Acros Organics
4-Aminophenyl ether, 98%
CAS: 101-80-4 | C12H12N2O | 200.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR104330050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Aminophenyl ether
CAS101-80-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0