Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips
Actual product may vary
Thermo Scientific
Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips
Combine these Thermo Scientific™ Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips with your pipette.
Have Questions?
Catalog NumberSterility
9420300Non-sterile
Catalog number 9420300
HSN 84799090
Price (MYR)
4,522.00
Pack
Sterility:
Non-sterile
Price (MYR)
4,522.00
Pack
Combine these Thermo Scientific™ Finnpipette™ Stepper Pipette Adapters for 25 and 50mL tips with your pipette. Choose sterile or nonsterile.
Specifications
Certifications/ComplianceClassification III (FL) according to DIN 12876-1
Height (Metric)62 cm
DescriptionFor 25 or 50mL Finnitip Stepper tips
IncludesRS232, SD memory card, USB, Ethernet, Modbus
Length (Metric)59 cm
Temperature (Metric) Maximum250°C
For Use WithFinnTip Stepper Tip (25, 50 mL)
Temperature Range (Metric)-30°C to +250°C
Width (Metric)25 cm
Dimensions (L x W x H)59 x 25 x 62 cm
SterilityNon-sterile
ModelPRESTO™ A30
Weight (Metric)62 kg
Display TypeTFT Touchscreen
TypeTemperature Control System
For Use With (Application)Ideal solution for highly precise, external temperature application