Finntip™ Pipette Tips
Actual product may vary
Finntip™ Pipette Tips
Ensure accurate, precise pipetting. Finntip™ Specific Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Have Questions?
Catalog NumberVolume (Metric)SterilityFit for PurposeColor Code
940217110 mLNon-sterileCertified QualityRed
Catalog number 9402171
HSN 84799090
Price (MYR)
138.00
PK
Volume (Metric):
10 mL
Sterility:
Non-sterile
Fit for Purpose:
Certified Quality
Color Code:
Red
Price (MYR)
138.00
PK
Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.
Key Features:
  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack
Proven quality
  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination
Versatile Tip Range
  • Volume range from 0.2 μL to 10 mL
  • 96-format volumes: 10, 250, 300 and 1000 μL
  • 384-format volumes: 20 and 50 μL
  • Macro volumes: 5 mL and 10 mL
Immediate traceability
  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options
Note:
Availability may vary by country.
Specifications
DescriptionFinntip 10mL, Non-Filtered, Non-sterile, Bulk
Volume (Metric)10 mL
FilteredNon-filtered
SterilityNon-sterile
ColorRed
ModelFinntip 10mL
Length (English)6 in.
Length (Metric)15cm
FormatBulk
Color CodeRed
For Use With (Equipment)Thermo Scientific Finnpipette
Product LineFinnTip
Fit for PurposeCertified Quality
Unit SizePK