NanoDrop™ Eight Spectrophotometer
Actual product may vary
Thermo Scientific
NanoDrop™ Eight Spectrophotometer
Avoid troubleshooting failed experiments with efficient analysis of your sample quantity and quality using the Thermo Scientifictm™ NanoDrop™ Eight UV-Vis spectrophotometer. Ideal for pharma/biotech and academic researchers measuring a larger number of nucleic acid and protein samples.
Have Questions?
Catalog NumberWavelength Range
912A1099190 to 850 nm
Catalog number 912A1099
HSN 90273010
Price (MYR)
176,891.40
Each
Wavelength Range:
190 to 850 nm
Price (MYR)
176,891.40
Each
Increase the chances of success in your analyses with efficient analysis of your sample quantity and quality using the Thermo Scientific™ NanoDrop™ Eight UV-Vis spectrophotometer. Ideal for pharma/biotech and academic researchers measuring larger number of nucleic acid and protein samples. NanoDrop Eight measures 8 samples at a time in less than 20 seconds. Software identifies sample contaminants and reports true sample concentration with the built-in Thermo Scientific™ Acclaro™ Sample Intelligence technology. Optional software allows you to more easily comply with 21 CFR Part 11 regulation.
Quantify your samples with confidence using the NanoDrop Eight Multichannel Spectrophotometer. This is the next generation NanoDrop mulitichannel instrument with improved hardware and the best software features from the Thermo Scientific™ NanoDrop™ One/OneC spectrophotometer.
Acclaro Contaminant Identification ensures samples are high quality.
Know the true concentration of your nucleic acid or protein without the interfering effects of sample contaminants. Understanding your sample composition helps downstream experiments go right the first time.
  • Contaminant identification – recognizes common contaminants in nucleic acid and protein samples that could inhibit downstream reactions or falsely elevate calculated concentrations
  • Corrected calculations – applies chemometric algorithms to determine the true concentration of your analyte without the contaminant
  • Differentiate DNA from RNA – algorithms can identify mammalian RNA contaminating a dsDNA sample or mammalian dsDNA contaminating an RNA sample
NanoDrop Eight spectrophotometer enhancements
  • Wider dynamic range – measure higher concentration samples without dilution – up to 10,000 ng/μL dsDNA or 145 mg/mL IgG
  • Auto-Measure and Auto-Blank functions – multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF
  • Ergonomic ambidextrous pipetting – Pedestal array is aligned perpendicular to the front of the instrument creating a natural pipetting motion for users; the pipette guide is magnetic and movable from side-to-side allowing left- and righthanded users to quickly and comfortably dispense samples
  • Automated exports for LIMS – set a directory for data to be automatically exported; have your LIMS software sweep the folder to pull in NanoDrop Eight instrument data for archiving
  • Regulatory compliance made easy
Optional Thermo Scientific™ SciVault™ Software helps your lab stay compliant with US FDA data regulations every step of the way.
  • Streamline management –SciVault Software integrates into NanoDrop Eight Software user interface
  • Comply with US FDA 21 CFR Part 11 Regulation – Control user account access, apply digital signatures, and review audit logs
  • Network multiple instruments – Install SciVault Software on a central computer to control privileges and view audit logs across multiple NanoDrop Eight Instruments in different labs
  • Validate your systems – IQ/OQ documents and services available to confirm your instrument is running within manufacturer’s specifications
  • Software updates to make your time more productive
 Ready to start – NanoDrop Eight Software comes hard-coded with the most frequently used measurement applications including dsDNA, RNA, Protein A280, and UV-Vis
Always improving – user input keeps NanoDrop Software constantly moving forward with updates that deliver greater capabilities and efficiencies in the lab. Visit thermoscientific.com/nanodropsw to see and download the latest software applications. Register your instrument there as well so you won’t miss any enhancements.
Specifications
Absorbance Accuracy3% at 0.97 A, 302 nm, 23 ± 2°C
Absorbance Range0.04 to 200 A (10 mm equivalent)
DescriptionNanoDrop Eight UV-Vis Spectrophotometer
Detection Limits2.0 to 10,000 ng/μL dsDNA; 0.03 to 145 mg/mL IgG
Detector Type2048-element CMOS Linear Image Sensor
Dimensions (L x W x H)24 x 33 x 17 cm (9.4 x 13 x 6.7 in.)
Light SourceXenon Flash
Wavelength Range190 to 850 nm
Electrical Requirements100/240 V
Measurement Time<20 sec. (for 8 samples)
Pathlength1.0 mm, 0.2 mm, 0.1 mm (automatically adjusting)
Power Consumption3 W at Idle; 15 W at Working Conditions
Sample Size1 to 2 μL
Spectral Resolution≤1.8 nm (FWHM at Hg 254 nm)
Warranty1 Year
Wavelength Accuracy± 1 nm
Weight (English)8.2 lb.
Weight (Metric)3.7 kg
Unit SizeEach