Block, 15 x 16 mm dia.
Actual product may vary
Thermo Scientific
Block, 15 x 16 mm dia.
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog NumberNo. of Places
8887010615
Catalog number 88870106
HSN 84199090
Price (MYR)
896.00
Each
No. of Places:
15
Price (MYR)
896.00
Each
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
  • Warranty: 2 years
Specifications
Description15 x 16mm dia. block
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds15 x 16mm
LightingLED
No. of Places15
TypeBlock
Dimensions (L x W x H)76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
Unit SizeEach