Sorvall™ Microtube Adapters
Sorvall™ Microtube Adapters
Actual product may vary
Sorvall™ Microtube Adapters
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ adapters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
76003759
also known as HER76003759
For 0.25/0.4mL microtubes
76003750
also known as 76003750FS
0.2mL microtubes
76003758
also known as 76003758FS
0.5/0.6mL microtubes
Catalog number 76003759
also known as HER76003759
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
For 0.25/0.4mL microtubes
Accommodate tubes from 0.2 to 0.6mL using Thermo Scientific™ Sorvall™ Microtube Adapters.
Ordering Information:
Sold in packs of 24
Specifications
For Use With (Equipment)For 0.25/0.4mL microtubes
DescriptionFor 0.25/0.4mL microtubes; Red
ColorRed
Unit SizeEA