Nalgene™ General Long-Term Storage Cryogenic Tubes
Nalgene™ General Long-Term Storage Cryogenic Tubes
Actual product may vary
Nalgene™ General Long-Term Storage Cryogenic Tubes
Nalgene™ General Long-Term Storage Cryogenic Tubes
Nalgene™ General Long-Term Storage Cryogenic Tubes
Nalgene™ General Long-Term Storage Cryogenic Tubes
Nalgene™ General Long-Term Storage Cryogenic Tubes
Enhance aseptic techniques with these externally threaded general long-term storage cryogenic tubes, ideal for research and general lab storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Sterility
5000-00202.0 mLSterile
5000-0050
also known as NLG5000-0050
5.0 mLSterile
Catalog number 5000-0020
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2.0 mL
Sterility:
Sterile
Enhance aseptic techniques with externally threaded Thermo Scientific™ Nalgene™ General Long-Term Storage Cryogenic Tubes. These storage tubes are ideally suited for research and general laboratory storage of high-value and genomic materials down to vapor phase liquid nitrogen temperatures.
  • Large writing areas accommodate more written sample information and graduations for easy visual volume assessment
  • Noncytotoxic raw materials for added sample security and tubes certified free of Dnase and RNase for genomic applications
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • System 100 Tubes offer a more dense storage option (10x10) to save precious storage space
  • Self standing tubes for ease of use on the benchtop
  • Blank, nonsterile and uncapped tubes available in bulk for less demanding applications
Specifications
Diameter (Metric)12.7 mm
Product TypeCryogenic Screw Top
DescriptionTube, sterile, 2.0 mL
Diameter (English) Outer0.5 in.
AutoclavableNot Autoclavable
Capacity (Metric)2.0 mL
Bottom ShapeConical
MaterialPolypropylene
Thread StyleExternal
Length (English)1.87 in.
No. per Case500/Cs.
No. per Pack25/Pk.
RNase FreeYes
Product LineNalgene™
SterilitySterile
GraduatedYes
Anti-RotationYes
Packaging Quantity500/Cs.
Closure MaterialPolyethylene
Capacity (English)0.0676 oz.
Dnase FreeYes
Closure TypeScrew Cap
PrintedYes
Unit SizePK