Nitric Acid, DILUT-IT Concentrate, 1.0M, J.T. Baker™
Nitric Acid, DILUT-IT Concentrate, 1.0M, J.T. Baker™
Nitric Acid, DILUT-IT Concentrate, 1.0M, J.T. Baker™
Nitric Acid, DILUT-IT Concentrate, 1.0M, J.T. Baker™
Nitric Acid, DILUT-IT Concentrate, 1.0M, J.T. Baker™
Prepare fresh, reliable standard solutions quickly and accurately. Nitric Acid, DILUT-IT Concentrate, 1.0M, J.T. Baker™ is a pre-mixed volumetric concentrate for dilution to 1L that is ideal for titrimetry and other forms of wet chemical analysis.
Have Questions?
Catalog NumberQuantityPackaging
4868
also known as GT4868
For dilution to 1 LLDPE ampule
Catalog number 4868
also known as GT4868
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
For dilution to 1 L
Packaging:
LDPE ampule
Request bulk or custom format
Specifications
Chemical Name or MaterialNitric acid
ColorColorless
CAS7697-37-2
Melting Point0°C
Boiling Point83°C
View more
DILUT-IT solutions are relied upon for fast, accurate results in life sciences and pharmaceutical quality control applications. Simply dilute the volumetric concentrate to 1 liter (carefully following instructions provided with the package) and the resulting solution will have an exact normality within ±0.1%.
  • Enhanced productivity: Eliminate expensive, time-consuming steps, such as the selection and analysis of starting reagents, and weighing and standardization of the final solution
  • Space saving: Products require only one-tenth of the space required for ready-made solutions
  • Reduced shipping costs: Spend less on shipping due to the reduced weight and package size of a concentrate