Nalgene™ Class B Polymethypentene (PMP) Volumetric Flasks with Screw Caps
Actual product may vary
Nalgene™ Class B Polymethypentene (PMP) Volumetric Flasks with Screw Caps
Nalgene™ Class B Polymethypentene (PMP) Volumetric Flasks with Screw Caps
Nalgene™ Class B Polymethypentene (PMP) Volumetric Flasks with Screw Caps
Thermo Scientific
Nalgene™ Class B Polymethypentene (PMP) Volumetric Flasks with Screw Caps
Protect against leakage, breakage, and contamination with these volumetric flasks, calibrated to contain and deliver at 20 degrees selsius
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeLimit of Error
4001-10001000 mL28 mm±0.60 mL
4001-0100100 mL20 mm±0.16 mL
Catalog number 4001-1000
HSN 39233090
Price (MYR)
2,463.00
Case
Capacity (Metric):
1000 mL
Closure Size:
28 mm
Limit of Error:
±0.60 mL
Price (MYR)
2,463.00
Case
Protect against leakage and breakage. Thermo Scientific™ Nalgene™ ASTM Class B Polymethylpentene (PMP) Volumetric Flasks with Screw Caps are calibrated To Contain and To Deliver at 20°C. Flasks are lighter in weight than glass, more transparent than polypropylene and offer excellent chemical resistance. Leakproof† protection when used with provided Nalgene closure.
  • Break resistance and excellent chemical resistance for years of dependable use in the lab
  • Individually hand calibrated, permanent engraved with a colored graduation line and oval marking spot for ease in reading fluid levels
  • No meniscus to confuse readings
  • Polypropylene closure
  • Meets ASTM™ E288 and ISO 384 Class B tolerances for consistent quality and dependability
  • Autoclavable/Graduated/Leakproof† when used with Nalgene closure
Warning: Autoclavable but accuracy will be affected. Completely disengage threads or remove cap before autoclaving.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.
Warning: Autoclavable but accuracy will be affected. Completely disengage threads or remove cap before autoclaving.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (English)32 oz.
Certifications/ComplianceMeets ASTM™ E288 and ISO 384 Class B tolerances for consistent quality and dependability
DescriptionFlask Volumetric PMP 1000mL
Capacity (Metric)1000 mL
MaterialPMP
Height (English)12.4 in.
Height (Metric)315 mm
Limit of Error±0.60 mL
No. per Case6
No. per Pack1
AutoclavableAutoclavable but will affect accuracy
DisposableReusable
GraduatedYes
Mouth StyleNarrow
SterilityNon-sterile
ClassB
Bottom ShapeRound
Closure TypeScrew
Diameter (Metric) Outer119 mm
Weight (Metric)110 g
Culture AreaNot applicable
Surface TreatmentNot applicable
Volume (Metric) Working1000 mL
Closure Size28 mm
Unit SizeCase