Search Fisher Scientific
| Catalog Number | Length (Metric) | Width (Metric) | Diameter (Metric) |
|---|---|---|---|
| 236015 | 300 m | 114 mm | 25.4 mm |
| 236013 | 300 m | 60 mm | 25.4 mm |
| 236014 | 300 m | 110 mm | 25.4 mm |
| 236016 | 300 m | 170 mm | 25.4 mm |
When printed on the appropriate Brady™ label, this ribbon gives excellent scratch and good chemical resistance.