Flash<i>Smart</i>&trade; Elemental Analyzer
Actual product may vary
FlashSmart™ Elemental Analyzer
Expand your CHNS/O analysis capabilities and simplify your workflow with an automated Elemental Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
11206155Elemental AnalyzerFlashSmart V CHNS/O (+ MVC)
11206100Elemental AnalyzerFlashSmart N/PROTEIN
11206105Elemental AnalyzerFlashSmart NC SOIL
11206110Elemental AnalyzerFlashSmart CHN
11206115Elemental AnalyzerFlashSmart CHNS
11206120Elemental AnalyzerFlashSmart CHN/O
11206125Elemental AnalyzerFlashSmart CHNS/O
11206130Elemental AnalyzerFlashSmart N ORG.
11206135Elemental AnalyzerFlashSmart NC ORG.
11206140Elemental AnalyzerFlashSmart NCS
11206145Elemental AnalyzerFlashSmart N LUBRICANT
11206150Elemental AnalyzerFlashSmart V CHN/O (+ MVC)
11206160Elemental AnalyzerFlashSmart V CHN/CHN (+ MVC)
11206165Elemental AnalyzerFlashSmart V CHNS/CHNS (+ MVC)
Catalog number 11206155
HSN 90278990
Price (MYR)
469,004.00
Each
Type:
Elemental Analyzer
Includes:
FlashSmart V CHNS/O (+ MVC)
Price (MYR)
469,004.00
Each
Solve more laboratory challenges, improve workflow, and minimize downtime when performing carbon, hydrogen, nitrogen, sulfur and oxygen (CHNS/O) analyses with the Thermo Scientific™ FlashSmart™ Elemental Analyzers. Based on the modified Dumas Method, the FlashSmart Elemental Analyzer is highly flexible, offering modularity that enables over 20 configurations in one instrument. Besides expanding your analytical capabilities, this analyzer features powerful software that supports automation and precise reports, making the system easier to use from start to finish.
FlashSmart Elemental Analyzer Features
Add automation to your daily organic elemental analysis for a wide range of applications without compromising precision and accuracy.
  • Simple design and usability
  • Improved CHNS/O workflow analysis
  • Higher sample throughput
  • Higher modularity
  • Dedicated features
Specifications
TypeElemental Analyzer
Detector TypeThermal Conductivity Detector
For Use With (Application)Organic Chemistry, Pharmaceuticals, Petro-Chemistry, Energy, Material Characterization
IncludesFlashSmart V CHNS/O (+ MVC)
Unit SizeEach