Vacuum Pump with UK, US, and Schuko Cables
Vacuum Pump with UK, US, and Schuko Cables
Actual product may vary
Welch
Vacuum Pump with UK, US, and Schuko Cables
Quiet in operation with very low vibration. the Fisherbrand™ Vacuum Pump is designed for general duty laboratory applications requiring only rough vacuum or moderate pressure.
Have Questions?
Catalog NumberPump Type
WEL411011-03Vacuum Pump
Catalog number WEL411011-03
Price (MYR)
-
Pump Type:
Vacuum Pump
  • Compact filtration pump with small footprint
  • Economical to purchase and operate
  • Quiet running oil free pump
  • Lightweight and highly portable
  • DN 6/8 hose nozzle For vacuum filtration.
Specifications
Max. Vacuum100 mbar
Electrical Requirements90/260 V 50/60 Hz with Schuko UK and US power leads
Dimensions (L x W x H)14.4 x 12.7 x 19.6 cm (5.6 x 5 x 7.7 in.)
Weight (English)2 lb.
Weight (Metric)2.3 kg
For Use WithSchuko, UK And US Vacuum Cables
Pump TypeVacuum Pump
Air Displacement15 L/min.
ApplicationsAspiration
Maximum Pressure2.5 bar
Unit SizeEach