U.S. Standard Brass and Stainless Steel Test Sieve Receivers
Actual product may vary
W.S. TYLER
U.S. Standard Brass and Stainless Steel Test Sieve Receivers
Fit all 8 and 12 in. diameter (20.3 and 30.5cm) sieves.
Have Questions?
Catalog NumberMaterial
TYL8493Stainless Steel
Catalog number TYL8493
Price (MYR)
742.00
Each
Material:
Stainless Steel
Price (MYR)
742.00
Each
Fit all 8 and 12 in. diameter (20.3 and 30.5cm) sieves.
Fit all 8 and 12 in. diameter (20.3 and 30.5cm) sieves.
Specifications
Depth (English)2 in.
Depth (Metric)5.08 cm
Product TypeFull Height Bottom Pan With Skirt
Diameter (English) Outer8 in.
MaterialStainless Steel
For Use With (Equipment)Test Sieve
Height (English)2.6 in.
Height (Metric)6.6 cm
Diameter (Metric) Outer203 mm
Mesh MaterialStainless Steel
Unit SizeEach