▸
Blood, Hematology and Coagulation Testing Products
▸
Blood Testing Reagents and Controls
▸
Hematology Controls
▸
Collagen Reagent
Actual product may vary
W.S. TYLER
Collagen Reagent
Collagen is used in routine platelet aggregation studies for evaluation of qualitative platelet function disorders.
Have Questions?
Catalog Number
Quantity
TYL5368
2 x 1 mL Vials
Catalog number
TYL5368
Price (MYR)
1,035.00
Each
Add to cart
Quantity:
2 x 1 mL Vials
Price (MYR)
1,035.00
Each
Add to cart
Collagen is used in routine platelet aggregation studies for evaluation of qualitative platelet function disorders.
Reagents are optimized for use with AggRAM, but may be used on any turbidometric aggregometer of similar quality
The collagen reagent is a clear, stabilized liquid reagent prepared from equine tendon, which is one of the worlds best collagen preparations
Collagen is used in routine platelet aggregation studies for evaluation of qualitative platelet function disorders
The collagen reagent stock solution id reconstituted to a concentration of 100 μg/mL
Specifications
Detectable Analytes
Platelets
For Use With (Equipment)
AggRAM
Sample Type
Plasma
Quantity
2 x 1 mL Vials
Form
Liquid
Unit Size
Each
Contents & Storage
2°C to 8°C