Glow-In-The-Dark/Ultra Reflective Letter: A
Actual product may vary
Thermo Scientific
Glow-In-The-Dark/Ultra Reflective Letter: A
Identify equipment, bins and areas of warehouse required by facility safety and maintenance. Brady™ Glow-In-The-Dark/Ultra Reflective Letter signs reflect light and glow brightly, combining a high level of reflectivity with advanced glow-in-the-dark technology.
Have Questions?
Catalog NumberSize (English)
TSS3020A3 x 6 in. (W x H)
Catalog number TSS3020A
Price (MYR)
3,439.00
Each
Size (English):
3 x 6 in. (W x H)
Price (MYR)
3,439.00
Each
  • Micro-prism reflectors for maximum visibility from greater distance, wider angle and lower light source
  • Designed for permanent Indoor/Outdoor applications
  • Made of high intensity prismatic reflective sheeting photoluminescent material with pressure sensitive adhesive
Specifications
Adhesive TypeAcrylic, Permanent
ColorBlack, Silver, Green
FinishReflective
FormatLabel
MaterialVinyl (B-309)
Product TypeUltra Reflective Letter Label
Display TextLegend: A
For Use With (Application)Labeling in Low Light Conditions (Indoor/Outdoor)
Size (English)3 x 6 in. (W x H)
Size (Metric)7.6 x 15.24cm
Width (English)3 in.
Height (English) Character5 in.
Unit SizeEach