TSE Series Glass Door Laboratory Refrigerators
Actual product may vary
TSE Series Glass Door Laboratory Refrigerators
TSE Series Glass Door Laboratory Refrigerators
TSE Series Glass Door Laboratory Refrigerators
TSE Series Glass Door Laboratory Refrigerators
TSE Series Glass Door Laboratory Refrigerators
Digital control, bright lighting, and temperature alarms deliver dependable storage for everyday supplies. Thermo Scientific TSE Series Glass Door Refrigerators provide clear visibility and simple inventory checks in convenient undercounter and upright single door sizes for routine lab use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Capacity (English)Electrical RequirementsStyle
TSE2205GV564 L20 cu.ft.230 V/50 HzUpright
TSE0405GV77 L3 cu.ft.230 V/50 HzUndercounter
TSE1205GV266 L9.4 cu.ft.230 V/50 HzUpright
Catalog number TSE2205GV
HSN 84184090
Price (MYR)
31,770.00
Each
Capacity (Metric):
564 L
Capacity (English):
20 cu.ft.
Electrical Requirements:
230 V/50 Hz
Style:
Upright
Price (MYR)
31,770.00
Each
Thermo Scientific TSE Series Glass Door Laboratory Refrigerators provide dependable temperature performance and straightforward operation for routine lab use. Bright visibility and digital control support easy access to everyday supplies, and all models use SNAP and F-Gas compliant hydrocarbon refrigerants. Designed to meet daily laboratory needs, TSE refrigerators include essential features and a two-year warranty.
  • Designed for everyday research and academic lab applications with good performance and easy-to-use digital controls
  • Quiet operation, less than 53 dB
  • SNAP compliant green refrigerants
  • Auto defrost
  • Four 2 in. casters, front casters are lockable
  • Bright LED interior lighting on glass door models
  • Hi/Lo Temperature, and door ajar failure visual alarms
  • Rear access port
  • Keyed door locks, compatible with popular E-lock adapters
Stacking Kit accessory available
Specifications
Capacity (English)20 cu.ft.
Capacity (Metric)564 L
Temperature Range1°C to 10°C
Temperature (Setpoint)5° C
RefrigerantR290
Door StyleGlass
No. of Doors1
Shelves4
Cabinet MaterialPainted Steel
Exterior FinishWhite-coated Steel
InsulationHFC-free Polyurethane Foam
Certifications/ComplianceCE
StyleUpright
DisplayDigital
CastersLocking
Control TypeDigital
Ports1 Rear-access Port
Warranty2 Years Parts & Labor
DefrostAuto
LightingLED
Monitoring OptionsAudible and Visual High and Low Temperature Alarms
Access SecurityKey Lock
CompressorHermetic
Dimensions (DxWxH) Exterior34.7 x 28.0 x 79.7 in. (88.1 x 71.3 x 202.4 cm)
Dimensions (D x W x H) Interior27.6 x 23.5 x 59.3 in. (70.0 x 59.6 x 150.5 cm)
Plug TypeUK
Electrical Requirements230 V/50 Hz
Energy Usage1.8 kw-hr/day
Shipping Weight (English)335 lb.
Shipping Weight (Metric)146 kg
Unit SizeEach