Specifications
Capacity (English)3 cu.ft.
Capacity (Metric)77 L
Temperature (Setpoint)-20°C
Door StyleSolid
No. of Doors1
Cabinet MaterialPainted Steel
Chamber MaterialPainted Steel
InsulationHFC-free Polyurethane Foam
Shelves2
Certifications/ComplianceCE
StyleUndercounter
DisplayDigital
CastersRear: Nylon Wheels
Control TypeDigital
Ports1 Rear-access Port
Warranty2 Years Parts and Labor
DefrostAuto
Dimensions (DxWxH) Exterior25.3 x 23.4 x 32.8 in. (64.5 x 59.5 x 83.3 cm)
Dimensions (D x W x H) Interior19.3 x 19.0 x 24.4 in. (49.0 x 48.3 x 62 cm)
Monitoring OptionsAlarm
Access SecurityKey Lock
Plug TypeUK
Electrical Requirements230 V/50 Hz
Energy Usage0.72 kw-hr/day
CompressorHermetic
RefrigerantR600a
Shipping Weight (English)115 lb.
Shipping Weight (Metric)52 kg
Unit SizeEach