CryoMed™ Controlled-Rate Freezer, General Purpose
Actual product may vary
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
CryoMed™ Controlled-Rate Freezer, General Purpose
Thermo Scientific
CryoMed™ Controlled-Rate Freezer, General Purpose
Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Electrical RequirementsModel
TSCM34XV34 L220-230 V, 50/60HzGeneral Purpose without Printer
TSCM17PV17 L220-230 V, 50/60HzGeneral Purpose with Printer
TSCM17XV17 L220-230 V, 50/60HzGeneral Purpose without Printer
Catalog number TSCM34XV
HSN 84184010
Price (MYR)
222,943.00
Each
Capacity (Metric):
34 L
Electrical Requirements:
220-230 V, 50/60Hz
Model:
General Purpose without Printer
Price (MYR)
222,943.00
Each
For customers in research, cell and gene therapy, vaccine production, and biobanking, the CryoMed Controlled-Rate Freezer (CRF) provides precise, repeatable freezing results that protect samples from intracellular freezing. The CryoMed CRF features enhanced data traceability via a touch screen user interface and offers customizable freezing profiles while supporting 21 CFR part 11 and GMP needs.
Thermo Scientific CryoMed Controlled-Rate Freezers (CRF) are designed with reliability, protection, and data traceability in mind for your sample preparation and GMP production needs.
User-friendly operation with enhanced data traceability
  • Intuitive touchscreen display allows for easy set-up, operation, and review of a freezing run
  • Six pre-set freezing profiles and space for up to 14 user-defined, 'custom' freeze profiles
  • Integrated touchscreen UI logs usage and events to support 21 CFR part 11 requirements
  • USB data export of .pdf file run logs support 21 CFR part 11
  • User security: 3 levels of user accessibility
  • 'Run last' feature allows the same profile to be run consecutively with the touch of a button
  • PC interface software provides remote operation capabilities, run review, and custom profile creation
Real-time run monitoring for sample protection
  • Chamber and sample temperatures are monitored by Type T thermocouples, eliminating lag time and providing 'real time' responsiveness
  • Standard alarms to alert users of thermocouple failures, heater malfunction, high/low temperature limits, temperature tracking, power failure, and completion of run
  • Remote alarm contacts
  • Optional thermal printer
Reliable temperature performance for high throughput usage
  • Dual solenoid valves are designed to balance LN2 injection volume for precise temperature control and accelerated freezing
  • Preventative maintenance indicator for LN2 solenoid replacement limits downtime
  • Temperature control and uniformity achieved via an air-handling and liquid nitrogen injection ring system
  • Compatible with VHP cleaning techniques
  • Environmentally friendly insulating foam
  • Type 304 stainless steel with exterior powder-coat finish
Warranty varies by region. Check with your local representative for more information
Specifications
TypeControlled-rate freezer
Capacity (English)1.2 cu. ft.
Capacity (Metric)34 L
Temperature Range-180°C to +50°C
No. of Doors1
Cabinet MaterialStainless Steel
Exterior FinishPowder-coated Steel
Certifications/ComplianceCE
StyleFreestanding
DisplayTouchscreen User Interface
WarrantyWarranty varies by region. Contact your local sales representative for more information
ModelGeneral Purpose without Printer
Sample LoadingFront-loading
Dimensions (DxWxH) Exterior24.3 x 43.3 x 21.7 in. (61 x 109.9 x 55.1 cm)
Dimensions (D x W x H) Interior13 x 13 x 12 in. (33 x 33 x 30.5 cm)
Monitoring OptionsAlarms, Remote Alarm Contacts
Data OutputsUSB data export
On-Board DataloggingYes
PC ProgrammabilityPC Interface Software Supported
ConnectivityThermo Fisher Connect compatible
Amperage5.5A
Plug TypeEU
Voltage208/230 V
Electrical Requirements220-230 V, 50/60Hz
Vial Capacity<800
CompressorAir-cooled
Weight (English)174 lb.
Weight (Metric)78.9 kg
Unit SizeEach