Range of columns that feature high levels of robustness, reproducibility and performance
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric) Inner
Film Thickness
Particle Size
SGE054613
60 m
0.22 mm
0.25 μm
0.25 μm
Catalog number SGE054613
Price (MYR)
30,340.00
EA
Add to cart
Length (Metric):
60 m
Diameter (Metric) Inner:
0.22 mm
Film Thickness:
0.25 μm
Particle Size:
0.25 μm
Price (MYR)
30,340.00
EA
Add to cart
Range of columns that feature high levels of robustness, reproducibility and performance
Low bleed; high inertness for most demanding analyses
High tolerance level to adverse conditions and samples
Manufactured to exact specifications, ensuring column-to-column reproducibilty
forte BPX-70 (70% Cyanopropyl Polysilphenylene-siloxane)—MS-premium, low bleed column with a polarity that has been designed for Fatty Acid Methyl ester (FAME) analysis