▸
Biochemical Reagents
▸
Peptides
▸
N-Acetyl-L-leucyl-L-leucyl-L-methional, Tocris Bioscience™
Actual product may vary
N-Acetyl-L-leucyl-L-leucyl-L-methional, Tocris Bioscience™
Cathepsin inhibitor
Have Questions?
Catalog Number
Quantity
SGE038410
10 mg
Catalog number
SGE038410
Price (MYR)
591.00
Pack
Add to cart
Quantity:
10 mg
Price (MYR)
591.00
Pack
Add to cart
Very potent inhibitor of cathepsin L (Ki = 0.6 nM) and the strongest inhibitor of cathepsin B (Ki = 100 nM) amongst the peptide aldehydes. Also inhibits processing of malaria aspartic hemoglobinases plasmepsins I and II
in vitro
.
Specifications
Purity
>98%
CAS
136632-32-1
Molecular Weight (g/mol)
401.566
Product Type
N-Acetyl-L-leucyl-L-leucyl-L-methional
Quantity
10 mg
Unit Size
Pack
Contents & Storage
Store at -20°C