PTFE Micro Magnetic Stir Bar
Actual product may vary
PTFE Micro Magnetic Stir Bar
PTFE Micro Magnetic Stir Bar
PTFE Micro Magnetic Stir Bar
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)
SGE00181010 mm3 mm
FKS1180889213 mm3 mm
FKS1265631620 mm3 mm
FKS1624585130 mm3 mm
FKS118889527 mm2 mm
Catalog number SGE001810
Price (MYR)
510.00
Each
Length (Metric):
10 mm
Diameter (Metric):
3 mm
Price (MYR)
510.00
Each
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Diameter (Metric)3 mm
Length (Metric)10 mm
Quantity10/Pk.
ShapeCylinder
TypeStir Bar, Micro
Unit SizeEach