PL6500 Lab Refrigerators
PL6500 Lab Refrigerators
Actual product may vary
PL6500 Lab Refrigerators
PL6500 Lab Refrigerators
PL6500 Lab Refrigerators
PL6500 Lab Refrigerators
Store general-purpose applications in Thermo Scientific™ PL6500 Lab Refrigerators, which feature advanced alarm options to meet the requirements of a laboratory environment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
PLR10061006 L
PLR386386 L
Catalog number PLR1006
Price (MYR)
21,000.00
Special offer
38,181.00
Save 17,181.00 (45%)
EA
Add to cart
Capacity (Metric):
1006 L
Price (MYR)
21,000.00
Special offer
38,181.00
Save 17,181.00 (45%)
EA
Add to cart
Store general-purpose applications in Thermo Scientific™ PL6500 Lab Refrigerators. All models feature advanced alarm options and reliable temperature stability to meet the demanding requirements of the laboratory environment.
*Available in China and limited Asia-Pacific regions only. Consult your representative for availability.
Temperature Control
  • Temperature range of 1° to 12°C (factory pre-set at 4°C)
  • Advanced air stability: ±3°C
  • Digital temperature controller with intuitive interface
  • High and low visual temperature and audio alarms
  • Automatic defrost
  • High-density, fluorine free insulation
  • Interior fan for forced air circulation (two fans on double-door model PLR1006)
Heavy-duty Construction
  • Adjustable shelves
  • Lockable, dual-pane glass doors
  • Standard casters
Other Features
  • Keyed ON/OFF switch
  • Interior lighting
  • Standard 25mm access port
Ordering Information:
Available in China and limited Asia-Pacific regions only. Consult your representative for availability.
Note:
For use in non-regulated environments.
  • Warranty varies by country, please contact us for details
  • Specifications
    Capacity (English)35.52 cu. ft.
    Capacity (Metric)1006 L
    Temperature Range1°C to 12°C
    Door StyleGlass
    Shelves12
    InsulationHigh-density, fluorine free insulation
    DefrostAutomatic
    Monitoring OptionsAlarm
    Depth (Metric) Interior111 cm
    Height (Metric) Exterior223 cm
    Height (Metric) Interior162 cm
    Length (Metric) Exterior80 cm
    Width (Metric) Exterior120 cm
    Width (Metric) Interior58 cm
    Plug TypeChina
    Voltage220 V
    Shipping Weight (Metric)238 kg
    Unit SizeEA