Muffle Furnace With Flap Door, 1200°C
Muffle Furnace With Flap Door, 1200°C
Actual product may vary
Muffle Furnace With Flap Door, 1200°C
Muffle Furnace With Flap Door, 1200°C
Nabertherm
Muffle Furnace With Flap Door, 1200°C
Muffle furnaces are the reliable and long-lasting all-rounders in the laboratory and are ideally suited for a large number of processes in the field of material research and heat treatment.
Have Questions?
Catalog NumberCapacity (Metric)Controller Type
NABL-094K2LN29 LB180
Catalog number NABL-094K2LN2
Price (MYR)
0.00
EA
Add to cart
Capacity (Metric):
9 L
Controller Type:
B180
Price (MYR)
0.00
EA
Add to cart
  • Optimal temperature uniformity of +/- 5K with closed fresh air inlet in empty workspace
  • Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Ceramic heating plates with integral heating elements which are safeguarded and easy to replace
  • Adjustable air inlet integrated in door
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
  • Defined application within the constraints of the operating instructions
  • Very short delivery times for standard furnaces and spare parts
Additional equipment
  • Chimney, chimney with fan or catalytic converter
  • Over-temperature limit controller with adjustable cutout temperature for thermal protection
  • Protective gas connection to purge with non-flammable gases
  • Manual or automatic gas supply system
  • Port for thermocouple in the rear wall or in the furnace door
  • Process control and documentation via VCD software package for monitoring, documentation and control
Specifications
Controller TypeB180
Capacity (Metric)9 L
Temperature (Metric)1200°C
TypeFurnace
MaterialTextured Stainless Steel
StyleFlap Door
Dimensions (D x W x H) Interior240 x 230 x 170 mm
Dimensions (L x W x H) Exterior455 x 415 x 515 mm
Width (Metric)415 mm
Width (Metric) Interior230 mm
Height (Metric)515 mm
Height (Metric) Interior170 mm
Depth (Metric)455 mm
Depth (Metric) Interior240 mm
Weight (Metric)35 kg
Wattage3.0 kW
Unit SizeEA