Lead-Free Cleanroom Autoclave Tape
Lead-Free Cleanroom Autoclave Tape
Actual product may vary
Micronova
Lead-Free Cleanroom Autoclave Tape
Lead-free autoclave indicator tape is a proven method of positively confirming completion of the sterilization cycle in both gravity discharge and vacuum autoclaves.
Have Questions?
Catalog NumberDimensions (LxW)
MICAUTOPC-3460 yds. x 0.75 in.
Catalog number MICAUTOPC-34
Price (MYR)
2,700.00
CS
Add to cart
Dimensions (LxW):
60 yds. x 0.75 in.
Price (MYR)
2,700.00
CS
Add to cart
Micronova Lead-Free Autoclave Indicator Tape is an easy to handle, saturated crepe paper tape with synthetic polyisoprene adhesive and green indicator lines that change to black when proper levels of steam pressure and temperature have been achieved.
  • Lead-free ink indicator lines cannot be rubbed off
  • Adheres on contact and stays in place through live steam pressure
  • For use in gravity discharge and vacuum autoclaves
  • Printed lines cannot be rubbed off and will not contaminate instruments
  • Removes quickly and easily
  • Easy to handle, Latex-free
  • Wound on a 3 in. plastic core for cleanroom use
  • Certifications: RoHS Compliant
Specifications
AutoclavableNot Autoclavable
Certifications/ComplianceRoHS Compliant; ISO 9001 Certified Facility; Certificate of Conformance
DescriptionGreen indicator lines change to black when proper levels of steam pressure & temperature are achieved
Adhesive TypeSynthetic
CoatingRubber, Synthetic Resins
FinishMatte
FormatRoll
MaterialSaturated Crepe Paper
Product TypeSteam Autoclave Tape
SterilitySterile
For Use With (Application)Indicator tape for steam sterilization cycles.
For Use With (Equipment)P-52, P-52W Tape Dispensers
Includes3 in. Plastic Core
Latex FreeYes
Quantity48/Cs.
Size (English)12 x 5 x 4 in.
Size (Metric)30.5 x 12.7 x 10.2 cm
Tensile Strength20 lbs./in.
Thickness (English)6.5 mil
Thickness (Metric)0.15 mm
Dimensions (LxW)60 yds. x 0.75 in.
Length (English)60 yd.
Length (Metric)55 m
Width (English)0.75 in.
Width (Metric)19.05 mm
Width (English) Tape0.75 in.
Width (Metric) Tape19.05 mm
Unit SizeCS