70% IPA is a general-purpose disinfectant for use in controlled environments. It is a blend containing 70% USP grade Isopropyl Alcohol (IPA) and 30% USP Water for Injection (WFI).
Have Questions?
Catalog Number
Form
LVA7030
Liquid
Catalog number LVA7030
Price (MYR)
-
Form:
Liquid
Bag-in-bottle trigger spray is an airtight closed system which maintains sterility and integrity of the content throughout use
The integrated collapsible bag eliminates the risk of airborne contamination and possibility of suck back during the use of conventional trigger spray
Cost effective - the system allows all content to be dispensed from the bottle to avoid any wastage
Double-bagged and double sealed polybag case liner
Guaranteed to have an endotoxin level of less than 0.25 EU/mL
Individually lot coded for ease of traceability and quality control